SY3H3D678ECB Synthonix Corporation

1-tert-Butyl 2-methyl piperidine-1,2-dicarboxylate

  • MDL number MFCD06795925
  • Cas Number : 167423-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Colorless - Yellow - Yellow-brown
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56278.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56278.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart