SY3H3D678ECF Synthonix Corporation

tert-Butyl 4-acetyl-2,2-dimethyloxazolidine-3-carboxylate

  • MDL number MFCD20232837
  • Cas Number : 433683-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56282.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56282.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart