SY3H3D678ED0 Synthonix Corporation

1-(Tert-butoxycarbonyl)-5,5-dimethylpyrrolidine-2-carboxylic acid

  • MDL number MFCD13190739
  • Cas Number : 900158-99-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56283.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56283.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart