SY3H3D678ED7 Synthonix Corporation

(2-((tert-Butyldimethylsilyl)oxy)phenyl)boronic acid

  • Cas Number : 929277-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/56294.html

Safety Information

MSDS https://synthonix.com/56294.html
Availability details for
Quantity
  • Availability
  • Add to Cart