SY3H3D678EDF Synthonix Corporation

8-(tert-Butoxy)-8-oxooctanoic acid

  • MDL number MFCD28505597
  • Cas Number : 234081-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Colorless to light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56308.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56308.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart