SY3H3D678EE2 Synthonix Corporation

tert-Butyl 3-propoxypyrrolidine-1-carboxylate

  • MDL number MFCD08064278
  • Cas Number : 1175835-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56312.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56312.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart