SY3H3D678EF2 Synthonix Corporation

tert-Butyl 4-(2-amino-1-hydroxyethyl)piperidine-1-carboxylate

  • Cas Number : 301221-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56338.html

Safety Information

MSDS https://synthonix.com/56338.html
Availability details for
Quantity
  • Availability
  • Add to Cart