SY3H3D678EF7 Synthonix Corporation

(S)-tert-Butyl 4-(2-aminopropyl)piperazine-1-carboxylate

  • Cas Number : 1017606-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56348.html

Safety Information

MSDS https://synthonix.com/56348.html
Availability details for
Quantity
  • Availability
  • Add to Cart