SY3H3D678EFC Synthonix Corporation

[2,2'-Bipyridine]-6,6'-dicarbonitrile

  • MDL number No MDL found
  • Cas Number : 4411-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56376.html

Safety Information

MSDS https://synthonix.com/56376.html
Availability details for
Quantity
  • Availability
  • Add to Cart