SY3H3D678F02 Synthonix Corporation

5-Bromo-1,10-phenanthroline hydrate

  • MDL number MFCD28384130
  • Cas Number : 855360-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56388.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56388.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart