SY3H3D678F09 Synthonix Corporation

2-Bromo-4'-chloro-1,1'-biphenyl

  • MDL number MFCD18072809
  • Cas Number : 179526-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Small lump / Colour: White - Pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56406.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56406.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart