SY3H3D678F0A Synthonix Corporation

4-Bromo-4'-Chloro-1,1'-biphenyl

  • Cas Number : 23055-77-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56407.html

Safety Information

MSDS https://synthonix.com/56407.html
Availability details for
Quantity
  • Availability
  • Add to Cart