SY3H3D678F11 Synthonix Corporation

3-Bromo-2-phenylimidazo[1,2-a]pyrimidine

  • MDL number MFCD06660438
  • Cas Number : 904814-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56426.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56426.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart