SY3H3D678F13 Synthonix Corporation

2-(4-Bromophenyl)-1H-imidazo[4,5-c]pyridine

  • Cas Number : 113270-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56428.html

Safety Information

MSDS https://synthonix.com/56428.html
Availability details for
Quantity
  • Availability
  • Add to Cart