SY3H3D678F21 Synthonix Corporation

2-Bromo-1-(6-bromonaphthalen-2-yl)ethanone

  • MDL number MFCD24186238
  • Cas Number : 50637-83-7
  • Molecular Weight : 328

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Yellow to brown powder or crystals
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/56476.html

Safety Information

signalword Warning
pcodes P305,P351,P338
MSDS https://synthonix.com/56476.html
pictograms GHS07
hstatements H302,H319
Availability details for
Quantity
  • Availability
  • Add to Cart