SY3H3D678F39 Synthonix Corporation

Benzyl azetidin-3-yl(methyl)carbamate hydrochloride

  • MDL number No MDL found
  • Cas Number : 1822874-05-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to light-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56614.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56614.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart