SY3H3D678F52 Synthonix Corporation

4-Bromo-4'-methyl-1,1'-biphenyl

  • MDL number MFCD00466236
  • Cas Number : 50670-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56694.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56694.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart