SY3H3D678F58 Synthonix Corporation

3-Benzyloxy-6-bromo-2-fluorophenylboronic acid

  • MDL number MFCD09475868
  • Cas Number : 957035-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56728.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56728.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart