SY3H3D678F59 Synthonix Corporation

(2-(Benzyloxy)-4-bromo-6-fluorophenyl)boronic acid

  • Cas Number : 1264175-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56729.html

Safety Information

MSDS https://synthonix.com/56729.html
Availability details for
Quantity
  • Availability
  • Add to Cart