SY3H3D678F5D Synthonix Corporation

(4-(Benzyloxy)-3,5-difluorophenyl)boronic acid

  • MDL number MFCD16295234
  • Cas Number : 156635-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56733.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56733.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart