SY3H3D678F81 Synthonix Corporation

tert-Butyl 3-bromo-1H-indole-1-carboxylate

  • MDL number MFCD05864772
  • Cas Number : 143259-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to Yellow or Pale-red Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56868.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56868.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart