SY3H3D678F84 Synthonix Corporation

2-(3-Bromo-2,2-dimethylpropyl)isoindoline-1,3-dione

  • Cas Number : 111992-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56871.html

Safety Information

MSDS https://synthonix.com/56871.html
Availability details for
Quantity
  • Availability
  • Add to Cart