SY3H3D678F89 Synthonix Corporation

tert-Butyl 4-(bromomethyl)-1H-benzo[d]imidazole-1-carboxylate

  • MDL number MFCD13182538
  • Cas Number : 132873-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56895.html

Safety Information

signalword Warning
pcodes P280
MSDS https://synthonix.com/56895.html
pictograms GHS07
hstatements H302, H315, H319, H332, H335
Availability details for
Quantity
  • Availability
  • Add to Cart