SY3H3D678F90 Synthonix Corporation

tert-Butyl (3-ethynylphenyl)carbamate

  • MDL number MFCD14585306
  • Cas Number : 185619-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56910.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56910.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart