SY3H3D678F92 Synthonix Corporation

tert-Butyl benzo[b]thiophen-2-ylcarbamate

  • MDL number MFCD22056377
  • Cas Number : 89673-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56916.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/56916.html
pictograms GHS07
hstatements H302, H315, H319, H332, H335
Availability details for
Quantity
  • Availability
  • Add to Cart