SY3H3D678FA5 Synthonix Corporation

7-(4-Bromobutoxy)-3,4-dihydroquinolin-2(1H)-one

  • MDL number MFCD06658540
  • Cas Number : 129722-34-5
  • Molecular Weight : 298.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/56958.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56958.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart