SY3H3D678FB9 Synthonix Corporation

1-Benzyl-5-methyl-1,2,3,6-tetrahydropyridin-4-ol

  • MDL number MFCD29760296
  • Cas Number : 2007909-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57020.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57020.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart