SY3H3D678FC3 Synthonix Corporation

tert-Butyl 6-hydroxyindoline-1-carboxylate

  • Cas Number : 957204-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57031.html

Safety Information

MSDS https://synthonix.com/57031.html
Availability details for
Quantity
  • Availability
  • Add to Cart