SY3H3D678FCA Synthonix Corporation

3-((tert-Butoxycarbonyl)amino)-4-methylbenzoic acid

  • MDL number MFCD01861378
  • Cas Number : 231958-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form White to Pale-purple or Yellow Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57044.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57044.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart