SY3H3D678FCB Synthonix Corporation

3-((tert-Butoxycarbonyl)amino)-4-methoxybenzoic acid

  • MDL number MFCD01765428
  • Cas Number : 306937-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to light-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57047.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57047.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart