SY3H3D678FD5 Synthonix Corporation

Benzyl 2-methylpiperazine-1-carboxylate

  • MDL number MFCD04115058
  • Cas Number : 444666-46-0
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Colorless to light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57071.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57071.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart