SY3H3D678FED Synthonix Corporation

tert-Butyl (1-(4-chlorophenyl)-2-hydroxyethyl)carbamate

  • MDL number MFCD24387088
  • Cas Number : 147353-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57105.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57105.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart