SY3H3D67900E Synthonix Corporation

(S)-2-(Butylsulfonamido)-3-(4-hydroxyphenyl)propanoic acid

  • MDL number MFCD07782139
  • Cas Number : 149490-60-8
  • Molecular Weight : 301.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57177.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57177.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart