SY3H3D679017 Synthonix Corporation

(S)-tert-Butyl (2-amino-1-phenylethyl)carbamate

  • MDL number MFCD09885895
  • Cas Number : 137102-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57197.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57197.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart