SY3H3D67901F Synthonix Corporation

tert-Butyl 2,2-difluoro-3-(methacryloyloxy)pentanoate

  • MDL number MFCD26743594
  • Cas Number : 1092693-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57217.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57217.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart