SY3H3D679028 Synthonix Corporation

tert-Butyl 1-oxo-2,6-diazaspiro[4.5]decane-6-carboxylate

  • MDL number MFCD15071621
  • Cas Number : 1221818-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57240.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57240.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart