SY3H3D67902B Synthonix Corporation

(S)-1-((S)-2-((tert-Butoxycarbonyl)amino)propanoyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD00134853
  • Cas Number : 33300-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white powder or ccrystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57243.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57243.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart