SY3H3D67902C Synthonix Corporation

tert-Butyl 4-propionylpiperidine-1-carboxylate

  • MDL number MFCD12912615
  • Cas Number : 419571-73-6
  • Molecular Weight : 241.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 4-propionyl-1-piperidinecarboxylate
Inchi Code 1S/C13H23NO3/c1-5-11(15)10-6-8-14(9-7-10)12(16)17-13(2,3)4/h10H,5-9H2,1-4H3
Inchi Key IGSFTWJIBCDRRD-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/57252.html

Safety Information

signalword Warning
pcodes P280,P305,P351,P338
MSDS https://synthonix.com/57252.html
pictograms GHS07
hstatements H317,H319
Availability details for
Quantity
  • Availability
  • Add to Cart