SY3H3D679031 Synthonix Corporation

(4R,5R)-3-tert-Butyl 4-methyl 2,2,5-trimethyloxazolidine-3,4-dicarboxylate

  • MDL number MFCD20488086
  • Cas Number : 1393440-06-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57258.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57258.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart