SY3H3D679032 Synthonix Corporation

(R)-4-(tert-Butoxy)-3-((tert-butoxycarbonyl)amino)-4-oxobutanoic acid

  • MDL number MFCD00236827
  • Cas Number : 77004-75-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57259.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57259.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart