SY3H3D679036 Synthonix Corporation

tert-Butyl 3-cyano-3-((trimethylsilyl)oxy)pyrrolidine-1-carboxylate

  • MDL number MFCD24466109
  • Cas Number : 942190-60-5
  • Molecular Weight : 284.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Colorless to light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57263.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57263.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart