SY3H3D67904B Synthonix Corporation

2-Bromo-9H-xanthen-9-one

  • Cas Number : 56341-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57302.html

Safety Information

MSDS https://synthonix.com/57302.html
Availability details for
Quantity
  • Availability
  • Add to Cart