SY3H3D679056 Synthonix Corporation

5-Bromo-1-phenyl-1H-benzo[d]imidazole

  • MDL number MFCD05864728
  • Cas Number : 221636-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Light-brown to brown or purple powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57340.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57340.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart