SY3H3D679057 Synthonix Corporation

6-Bromo-2-phenylimidazo[1,2-a]pyridine

  • MDL number MFCD00219253
  • Cas Number : 4044-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57341.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57341.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart