SY3H3D67907A Synthonix Corporation

2,2'-Bis(trifluoromethyl)-[1,1'-biphenyl]-4,4'-diamine

  • MDL number No MDL found
  • Cas Number : 341-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57468.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57468.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart