SY3H3D67907E Synthonix Corporation

4-(4-Bromo-3-(hydroxymethyl)phenoxy)benzonitrile

  • Cas Number : 906673-45-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57484.html

Safety Information

MSDS https://synthonix.com/57484.html
Availability details for
Quantity
  • Availability
  • Add to Cart