SY3H3D679080 Synthonix Corporation

1,2-Bis(4-fluorophenyl)-2-hydroxyethanone

  • Cas Number : 53458-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57494.html

Safety Information

MSDS https://synthonix.com/57494.html
Availability details for
Quantity
  • Availability
  • Add to Cart