SY3H3D67909E Synthonix Corporation

(2-Bromophenyl)(2,4-dimethylphenyl)sulfane

  • MDL number MFCD26386449
  • Cas Number : 960203-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Yellow liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57588.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57588.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart