SY3H3D6790AD Synthonix Corporation

4,4'-Bis(hydroxymethyl)biphenyl

  • MDL number MFCD00016713
  • Cas Number : 1667-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57654.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57654.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart