SY3H3D6790C6 Synthonix Corporation

tert-Butyl 3-iodo-5-methoxy-1H-indole-1-carboxylate

  • MDL number MFCD05864763
  • Cas Number : 192189-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form White to light-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57717.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57717.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart